Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSGFSTEER AAPFSLEYRV FLKNEKGQYI SPFHDIPIYA DKDVFHMVVE VPRWSNAKME IATKDPLNPI KQDVKKGKLR YVANLFPYKG YIWNYGAIPQ TWEDPGHNDK HTGCCGDNDP IDVCEIGSKV CARGEIIGVK VLGILAMIDE GETDWKVIAI NVDDPDAANY NDINDVKRLK PGYLEATVDW FRRYKVPDGK PENEFAFNAE FKDKDFAIDI IKSTHDHWKA LVTKKTNGKG ISCMNTTLSE SPFKCDPDAA RAIVDALPPP CESACTVPTD VDKWFHHQKN (1-289 aa encoded by BC001022)
The protein encoded by this gene is a member of the inorganic pyrophosphatase (PPase) family. PPases catalyze the hydrolysis of pyrophosphate to inorganic phosphate, which is important for the phosphate metabolism of cells. Studies of a similar protein in bovine suggested a cytoplasmic localization of this enzyme.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytosolic inorganic pyrophosphatase; diphosphate phosphohydrolase; epididymis secretory sperm binding protein Li 66p; inorganic diphosphatase; inorganic diphosphatase 1; Inorganic pyrophosphatase; inorganic pyrophosphatase 1; PPase; pyrophosphatase 1; Pyrophosphate phospho-hydrolase; RP11-367H5.1
Gene Aliases: 2010317E03Rik; HEL-S-66p; IOPPP; PP; PP1; PPA1; Pyp; SID6-8061
UniProt ID: (Human) Q15181, (Mouse) Q9D819
Entrez Gene ID: (Human) 5464, (Rat) 294504, (Mouse) 67895
Molecular Function:
pyrophosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support