Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Immunogen sequence: MSGFSTEERA APFSLEYRVF LKNEKGQYIS PFHDIPIYAD KDVFHMVVEV PRWSNAKMEI ATKDPLNPIK QDVKKGKLRY VANLFPYKGY IWNYGAIPQT WEDPGHNDKH TGCCGDNDPI DVCEIGSKVC ARGEIIGVKV LGILAMIDEG ETDWKVIAIN VDDPDAANYN DINDVKRLKP GYLEATVDWF RRYKVPDGKP ENEFAFNAEF KDKDFAIDII KSTHDHWKAL VTKKTNGKGI SCMNTTLSES PFKCDPDAAR AIVDALPPPC ESACTVPTDV DKWFHHQKN
The protein encoded by this gene is a member of the inorganic pyrophosphatase (PPase) family. PPases catalyze the hydrolysis of pyrophosphate to inorganic phosphate, which is important for the phosphate metabolism of cells. Studies of a similar protein in bovine suggested a cytoplasmic localization of this enzyme.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytosolic inorganic pyrophosphatase; diphosphate phosphohydrolase; epididymis secretory sperm binding protein Li 66p; inorganic diphosphatase; inorganic diphosphatase 1; Inorganic pyrophosphatase; inorganic pyrophosphatase 1; PPase; pyrophosphatase 1; Pyrophosphate phospho-hydrolase; RP11-367H5.1
Gene Aliases: HEL-S-66p; IOPPP; PP; PP1; PPA1; Pyp; SID6-8061
UniProt ID: (Human) Q15181
Entrez Gene ID: (Human) 5464, (Rat) 294504
Molecular Function:
pyrophosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support