Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSALLRLLRT GAPAAACLRL GTSAGTGSRR AMALYHTEER GQPCSQNYRL FFKNVTGHYI SPFHDIPLKV NSKEENGIPM KKARNDEYEN LFNMIVEIPR WTNAKMEIAT KEPMNPIKQY VKDGKLRYVA NIFPYKGYIW NYGTLPQTWE DPHEKDKSTN CFGDNDPIDV CEIGSKILSC GEVIHVKILG ILALIDEGET DWKLIAINAN DPEASKFHDI DDVKKFKPGY LEATLNWFRL YKVPDGKPEN QFAFNGEFKN KAFALEVIKS THQCWKALLM KKCNGGAINC TNVQISDSPF RCTQEEARSL VESVSSSPNK ESNEEEQVWH FLGK
Hydrolyzes inorganic pyrophosphate. This activity is essential for correct regulation of mitochondrial membrane potential, and mitochondrial organization and function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20459; Inorganic pyrophosphatase 2, mitochondrial; PPase 2; Pyrophosphatase SID6-306; Pyrophosphate phospho-hydrolase 2
Gene Aliases: HSPC124; PPA2; RGD1307965; SID6-306
UniProt ID: (Human) Q9H2U2
Entrez Gene ID: (Human) 27068, (Rat) 310856
Molecular Function:
pyrophosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support