Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
This affinity purified antibody is directed against human PPAR gamma protein. A BLAST analysis was used to suggest broad reactivity with this protein from several sources. Expect reactivity against human (g1 and g2), mouse (g1 and g2), rat, squirrel, orangutan, duck (g1), macaque (g1-g7), boar (g1a-1d), mink, guinea pig, rabbit (g1 and g3), dog and hamster sources based on 100% homology for the immunogen sequence. Cross reactivity with PPAR gamma homologues from other sources has not been determined. No reactivity is expected against other subtypes of PPAR.
This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) subfamily of nuclear receptors. PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate transcription of various genes. Three subtypes of PPARs are known: PPAR-alpha, PPAR-delta, and PPAR-gamma. The protein encoded by this gene is PPAR-gamma and is a regulator of adipocyte differentiation. Additionally, PPAR-gamma has been implicated in the pathology of numerous diseases including obesity, diabetes, atherosclerosis and cancer. Alternatively spliced transcript variants that encode different isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HUMPPARG; nuclear hormone receptor; nuclear hormone receptor; PPARG; Nuclear receptor subfamily 1 group C member 3; nuclear receptor transcription factor; OTTHUMP00000185032; OTTHUMP00000185036; peroxisome proliferative activated receptor, gamma; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activated receptor gamma; peroxisome proliferator activated receptor gamma 2; peroxisome proliferator activated receptor gamma 4; peroxisome proliferator activated receptor gamma-1; peroxisome proliferator activatived receptor gamma; peroxisome proliferator activator receptor, gamma; peroxisome proliferator-activated nuclear receptor gamma variant 1; Peroxisome proliferator-activated receptor gamma; peroxisome proliferator-activated receptor gamma 1; peroxisome proliferator-activated receptor gamma 1-a; peroxisome proliferator-activated receptor gamma 1-b; peroxisome proliferator-activated receptor gamma 2; PPAR gamma; PPAR gamma 1; PPAR gamma 3; PPAR-gamma; PPARgamma; PPARy; transcription factor
Gene Aliases: CIMT1; GLM1; NR1C3; PPAR-GAMMA; PPAR-gamma2; PPARG; PPARG1; PPARG2; PPARGAMMA; PPARgamma2
UniProt ID: (Human) P37231, (Pig) O62807, (Dog) Q4U3Q4, (Rhesus monkey) O18924, (Chinese hamster) P57797, (Mouse) P37238, (Rat) O88275
Entrez Gene ID: (Human) 5468, (Pig) 397671, (Dog) 403606, (Rhesus monkey) 574190, (Rabbit) 100008892, (Chinese hamster) 100689245, (Guinea pig) 100729138, (Mouse) 19016, (Rat) 25664
Molecular Function: C4 zinc finger nuclear receptor DNA-binding transcription factor zinc finger transcription factor gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support