Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GIGHTTGKP LHFKGCPFHR IIKKFMIQGG DFSNQNGTGG ESIYGEKFED ENFHYKHDRE GLLSMANAGR NTNGSQFFIT TVPTPHLDGK HVVFGQVIKG IGVARILENV EVKGEKPAKL CVIAECGELK EGDDGGIFPK DGSGDSHPDF PEDADIDLKD VDKILLITED LKNIGNTFFK SQNWEMAIKK YAEVLRYVDS SKAVIETADR AKLQPIALSC VLNIGACKLK MSNWQGAIDS CLEALELDPS NTKALYRRAQ GWQGLKEYDQ ALADLKKAQG IAPEDKAIQA ELLKVKQKIK AQKDKEKAVY AKMFA (57-370 aa encoded by B C030707)
This gene encodes a H1 histone binding protein that is involved in transporting histones into the nucleus of dividing cells. Multiple isoforms are encoded by transcript variants of this gene. The somatic form is expressed in all mitotic cells, is localized to the nucleus, and is coupled to the cell cycle. The testicular form is expressed in embryonic tissues, tumor cells, and the testis. In male germ cells, this protein is localized to the cytoplasm of primary spermatocytes, the nucleus of spermatids, and the periacrosomal region of mature spermatozoa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40 kDa peptidyl-prolyl cis-trans isomerase; 40 kDa peptidyl-prolyl cis-trans isomerase D; cyclophilin 40; Cyclophilin D; Cyclophilin-40; Cyclophilin-related protein; Cyp 40; Cyp D; CYP-40; cytoplasmic cyclophilin D; EC 5.2.1.8; MGC33096; Peptidyl-prolyl cis-trans isomerase D; PPIase D; rotamase; Rotamase D; testicular tissue protein Li 147
Gene Aliases: 4930564J03Rik; CYP-40; CYP40; CYPD; PPID; Ppidl; Ppif
UniProt ID: (Human) Q08752, (Mouse) Q9CR16, (Rat) Q6DGG0
Entrez Gene ID: (Human) 5481, (Mouse) 67738, (Rat) 361967
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support