Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGIKVQRPR CFFDIAINNQ PAGRVVFELF SDVCPKTCEN FRCLCTGEKG TGKSTQKPLH YKSCLFHRVV KDFMVQGGDF SEGNGRGGES IYGGFFEDES FAVKHNKEFL LSMANRGKDT NGSQFFITTK PTPHLDGHHV VFGQVISGQE VVREIENQKT DAASKPFAEV RILSCGELIP KSKVKKEEKK RHKSSSSSSS SSSDSDSSSD SQSSSDSSDS ESATEEKSKK RKKKHRKNSR KHKKEKKKRK KSKKSASSES EAENLEAQPQ STVRPEEIPP IPENRFLMRK SPPKADEKER KNRERERERE CNPPNSQPAS YQRRLLVTRS GRKIKGRGPR VGDSFPRDLH NIAFVF (1-355 aa encoded by B C001555)
Structural Maintenance of Chromosomes (SMC) family proteins play critical roles in various nuclear events that require structural changes of chromosomes, including mitotic chromosome organization, DNA recombination and repair and global transcriptional repression. The chromo proteins are conserved in eukaryotes and lead to mitotic chromosome segregation defects. This suggests a critical function of SMC family proteins in mitotic chromosome dynamics. SMC1 and SMC3 form a heterodimeric complex required for metaphase progression in mitotic cells. Specifically this SMC1/SMC3 complex is responsible for sister chromatid cohesion during metaphase. Mutations in the gene can lead to Cornelia de Lange syndrome 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CARS-cyclophilin; CARS-Cyp; CASP10; Clk-associating RS-cyclophilin; Cyclophilin G; MGC133241; OTTHUMP00000204761; OTTHUMP00000204762; Peptidyl-prolyl cis-trans isomerase G; peptidyl-prolyl isomerase G (cyclophilin G); peptidylprolyl isomerase G; peptidylprolyl isomerase G (cyclophilin G); PPIase G; Rotamase G; SR-cyclophilin; SR-cyp; SR-related CTD-associated factor 10
Gene Aliases: AU019516; AU022200; B230312B02Rik; CARS-Cyp; CYP; PPIG; SCAF10; SRCyp
UniProt ID: (Human) Q13427, (Mouse) A2AR02
Entrez Gene ID: (Human) 9360, (Mouse) 228005
Molecular Function:
chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support