Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSALEKSMH LGRLPSRPPL PGSGGSQSGA KMRMGPGRKR DFSPVPWSQY FESMEDVEVE NETGKDTFRV YKSGSEGPVL LLLHGGGHSA LSWAVFTAAI ISRVQCRIVA LDLRSHGETK VKNPEDLSAE TMAKDVGNVV EAMYGDLPPP IMLIGHSMGG AIAVHTASSN LVPSLLGLCM IDVVEGTAMD ALNSMQNFLR GRPKTFKSLE NAIEWSVKSG QIRNLESARV SMVGQVKQCE GITSPEGSKS IVEGIIEEEE EDEEGSESIS KRKKEDDMET KKDHPYTWRI ELAKTEKYWD GWFRGLSNLF LSCPIPKLLL LAGVDRLDKD LTIGQMQGKF QMQVLPQCGH AVHEDAPDKV AEAVATFLIR HRFAEPIGGF QCVFPGC (1-386 aa encoded by BC050705)
Protein phosphatase methylesterase-1 catalyzes the demethylation of the protein phosphatase-2A catalytic subunit.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PME-1; Protein phosphatase methylesterase 1; protein phosphatase methylesterase-1; testicular secretory protein Li 39
Gene Aliases: 1110069N17Rik; 2700017M01Rik; PME-1; PME1; PP2593; PPME1; PRO0750; RGD1309683
UniProt ID: (Human) Q9Y570, (Mouse) Q8BVQ5, (Rat) Q4FZT2
Entrez Gene ID: (Human) 51400, (Mouse) 72590, (Rat) 361613
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support