Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IDIDDLLDAD SDEERASKLQ EALVDCY
Antigen sequence identity: Mouse 93%; Rat 96%
Inhibitor of the PP1 regulatory subunit PPP1CA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kinase C-enhanced PP1 inhibitor; Kinase-enhanced PP1 inhibitor; PKC-potentiated PP1 inhibitory protein; Protein phosphatase 1 regulatory subunit 14C; protein phosphatase 1, regulatory (inhibitor) subunit 14C; Serologically defined breast cancer antigen NY-BR-81
Gene Aliases: CPI17-like; KEPI; NY-BR-81; PPP1R14C
UniProt ID: (Human) Q8TAE6
Entrez Gene ID: (Human) 81706
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support