Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PP1R14C Polyclonal Antibody detects endogenous levels of PP1R14C protein.
Inhibitor of the PP1 regulatory subunit PPP1CA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kinase C-enhanced PP1 inhibitor; Kinase-enhanced PP1 inhibitor; PKC-potentiated PP1 inhibitory protein; Protein phosphatase 1 regulatory subunit 14C; protein phosphatase 1, regulatory (inhibitor) subunit 14C; Serologically defined breast cancer antigen NY-BR-81
Gene Aliases: 6330514J04Rik; CPI17-like; KEPI; NY-BR-81; PPP1R14C
UniProt ID: (Human) Q8TAE6, (Mouse) Q8R4S0, (Rat) Q8R4R9
Entrez Gene ID: (Human) 81706, (Mouse) 76142, (Rat) 171010
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support