Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VFQPPPPPPP APSGDAPAEK ERCPAGQQPP QHRVAPARPP PKPREVRKIK PVEVEESGES EAESAPGEVI QAEVTVHAAL ENGSTVATAA SPAPEEPKAQ A
interacts with protein phosphatase 1 and actin filaments; involved in regulation of synaptic transmission.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: neurabin II; Neurabin-2; Neurabin-II; Protein phosphatase 1 regulatory subunit 9B; protein phosphatase 1, regulatory (inhibitor) subunit 9B; protein phosphatase 1, regulatory subunit 9B; protein phosphatase 1, regulatory subunit 9B, spinophilin; Spinophilin
Gene Aliases: PPP1R6; PPP1R9; PPP1R9B; SPINO; SPL; Spn
UniProt ID: (Human) Q96SB3, (Mouse) Q6R891
Entrez Gene ID: (Human) 84687, (Mouse) 217124
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support