Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
PPP2R2D is the B regulatory subunit might modulate substrate selectivity and catalytic activity, and also might direct the localization of the catalytic enzyme to a particular subcellular compartment.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PP2A subunit B isoform B55-delta; PP2A subunit B isoform delta; PP2A subunit B isoform PR55-delta; PP2A subunit B isoform R2-delta; protein phosphatase 2 regulatory subunit B, delta; protein phosphatase 2, regulatory subunit B, delta isoform; Serine/threonine-protein phosphatase 2A 55 kDa regulatory subunit B delta isoform
Gene Aliases: KIAA1541; MDS026; PPP2R2D
UniProt ID: (Human) Q66LE6
Entrez Gene ID: (Human) 55844
Molecular Function:
protein phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support