Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LATPNTCPNKK KSEQELKDEE MDLFTKYYSE WKGGRKNTNE FYKTIPRFYY RLPAEDEVLL QKLREESRAV FLQRKSRELL DNEELQNLWF LLDKHQTPPM IGEEAMINYE NF
Highest antigen sequence identity to the following orthologs - mouse 96%, rat 95%.
P2R3C may regulate MCM3AP phosphorylation through phosphatase recruitment. It may play a role in the activation-induced cell death of B-cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: protein phosphatase 2 (formerly 2A), regulatory subunit B''; protein phosphatase 2 regulatory subunit B'', gamma; protein phosphatase 2, regulatory subunit B'', gamma; Protein phosphatase subunit G5PR; rhabdomyosarcoma antigen MU-RMS-40.6A; Rhabdomyosarcoma antigen MU-RMS-40.6A/6C; rhabdomyosarcoma antigen Mu-RMS-40.6C; Serine/threonine-protein phosphatase 2A regulatory subunit B'' subunit gamma
Gene Aliases: C14orf10; G4-1; G5PR; PPP2R3C
UniProt ID: (Human) Q969Q6
Entrez Gene ID: (Human) 55012
Molecular Function:
protein phosphatase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support