Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PRCPGESSHI CDFIRKTLNA GAYSKVVQER LVQAEYWHDP IKEDVYRNHS IFLADINQER GINESYKKNL MALKKFVMVK FLNDSIVDPV DSEWFGFYRS GQAKETIPLQ ETSLYTQDRL GLKEMDNAGQ LVFLATEGDH LQLSEEWFYA HIIPFLG
The protein encoded by this gene is a small glycoprotein involved in the catabolism of lipid-modified proteins during lysosomal degradation. The encoded enzyme removes thioester-linked fatty acyl groups such as palmitate from cysteine residues. Defects in this gene are a cause of infantile neuronal ceroid lipofuscinosis 1 (CLN1, or INCL) and neuronal ceroid lipofuscinosis 4 (CLN4). Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3.1.2.22; ceroid-palmitoyl-palmitoyl-protein thioesterase 1; Palmitoyl-protein hydrolase 1; Palmitoyl-protein thioesterase 1; PPT-1
Gene Aliases: 9530043G02Rik; AA960502; C77813; CLN1; D4Ertd184e; INCL; PPT; PPT1
UniProt ID: (Human) P50897, (Rat) P45479, (Mouse) O88531
Entrez Gene ID: (Human) 5538, (Rat) 29411, (Mouse) 19063
Molecular Function:
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support