Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Control: human PPY-transfected 293T cells
Predicted Reactivity: Rhesus Monkey (100%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Pancreatic polypeptide (PP) is a 36 amino-acid residue peptide with sequence and 3-D structural homology with NPY and PYY. PP is the least conserved of the three peptides and was the first to be identified. It is almost exclusively expressed in the endocrine pancreas and released in response to cholinergic stimulation and food intake. PP has inhibitory effects on gall bladder secretion, intestinal motility and pancreatic secretion. PP may serve as a marker for certain gut endocrine tumors including carcinoid tumors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGNC:9327; IOPPP; Pancreatic polypeptide prohormone; Pancreatic polypeptide Y; PH; PPA1; prepro-PP (prepropancreatic polypeptide); SID6-8061
Gene Aliases: PNP; PP; PPY
UniProt ID: (Human) P01298
Entrez Gene ID: (Human) 5539
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support