Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: HeLa, MCF7, NIH/3T3, RD. The target is usually found in the following locations: Cytoplasm, cytosol.
Immunogen sequence: STDDGSLSEE TPAGPPTCSV PPASALPTQQ YAKSLPVSVP VWGFKEKRTE ARSSDEENGP PSSPDLDRIA ASMRALVLRE AEDTQVFGDL PRPRLNTSDF QKLKRKY
The Akt signaling pathway contributes to the regulation of apoptosis after a variety of cell death signals. AKT1S1, also known as PRAS40, is a proline-rich substrate of the kinase AKT1 and is thought to play a role in neuroprotection mediated by nerve growth factor (NGF) after transient focal cerebral ischemia. AKT1S1 is also a substrate and potential regulator of mammalian target of rapamycin (mTOR), a serine/threonine kinase that regulates cell growth and cell cycle, and a negative regulator of autophagy. Treatment with the insulin-like growth factor-1 (IGF1) can induce the phosphorylation of AKT1S1 via the PI3K/AKT signaling pathway in PC12 cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 40 kDa proline-rich AKT substrate; AKT1 substrate 1 (proline rich); AKT1 substrate 1 (proline-rich); AKT1S1; OTTHUMP00000196756; OTTHUMP00000196760; Proline-rich AKT substrate; Proline-rich AKT1 substrate 1
Gene Aliases: 1110012J22Rik; AI227026; AKT1S1; Lobe; Lobel; Pras; PRAS40
UniProt ID: (Human) Q96B36, (Mouse) Q9D1F4
Entrez Gene ID: (Human) 84335, (Mouse) 67605
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support