Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MRRSEVLAEE SIVCLQKALN HLREIWELIG IPEDQRLQRT EVVKKHIKEL LDMMIAEEES LKERLIKSIS VCQKELNTLC SELHVEPFQE EGETTILQLE KDLRTQVELM RKQKKERKQE LKLLQEQDQE LCEILCMPHY DIDSASVPSL EELNQFRQHV TTLRETKASR REEFVSIKRQ IILCMEELDH TPDTSFERDV VCEDEDAFCL SLENIATLQK LLRQLEMQKS QNEAVCEGLR TQIRELWDRL QIPEEEREAV ATIMSGSKAK VRKALQLEVD RLEELKMQNM KKVIEAIRVE LVQYWDQCFY SQEQRQAFAP FCAEDYTESL LQLHDAEIVR LKNYYEVHKE LFEGVQKWEE TWRLFLEFER KASDPNRFTN RGGNLLKEEK QRAKLQKMLP KLEEELKARI ELWEQEHSKA FMVNGQKFME YVAEQWEMHR LEKERAKQER QLKNKKQTET EMLYGSAPRT PSKRRGLAPN TPGKARKLNT TTMSNATANS SIRPIFGGTV YHSPVSRLPP SGSKPVAAST CSGKKTPRTG RHGANKENLE LNGSILSGGY PGSAPLQRNF SINSVASTYS EFAKDPSLSD SSTVGLQREL SKASKSDATS GILNSTNIQS
This gene encodes a protein that is involved in cytokinesis. The protein is present at high levels during the S and G2/M phases of mitosis but its levels drop dramatically when the cell exits mitosis and enters the G1 phase. It is located in the nucleus during interphase, becomes associated with mitotic spindles in a highly dynamic manner during mitosis, and localizes to the cell mid-body during cytokinesis. This protein has been shown to be a substrate of several cyclin-dependent kinases (CDKs). It is necessary for polarizing parallel microtubules and concentrating the factors responsible for contractile ring assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: anaphase spindle elongation 1 homolog; MGC1671; MGC3669; PRC1; protein regulating cytokinesis 1; Protein regulator of cytokinesis 1
Gene Aliases: ASE1; PRC1
UniProt ID: (Human) O43663
Entrez Gene ID: (Human) 9055
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support