Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SEQALADFA ELIKHLKRTI PGAENQPVIA IGGSYGGMLA AWFRMKYPHM VVGALAASAP IWQFEDLVPC GVFMKIVTTD FRKSGPHCSE SIHRSWDAIN RLSNTGSGLQ WLTGALHLCS PLTSQDIQHL KDWISETWVN LAMVDYPYAS NFLQPLPAWP IKVVCQYLKN PNVSDSLLLQ NIFQALNVYY NYSGQVKCLN ISETATSSLG TLGWSYQACT EVVMPFCTNG VDDMFEPHSW NLKELSDDCF QQWGVRPRPS WITTMYGGKN ISSHTNIVFS NGELDPWSGG GVTKDITDTL VAVTISEGAH HLDLRTKNAL DPMSVLLARS LEVRHMKNWI RDFYDSAGKQ H (147-496 aa encoded by BC001500)
Cleaves C-terminal amino acids linked to proline in peptides such as angiotensin II, III and des-Arg9-bradykinin. This cleavage occurs at acidic pH, but enzymatic activity is retained with some substrates at neutral pH.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Angiotensinase C; Lysosomal carboxypeptidase C; Lysosomal Pro-X carboxypeptidase; PRCP; Proline carboxypeptidase; Prolylcarboxypeptidase; prolylcarboxypeptidase (angiotensinase C)
Gene Aliases: 2510048K03Rik; 2610104A14Rik; AI451719; HUMPCP; PCP; PRCP
UniProt ID: (Human) P42785, (Mouse) Q7TMR0
Entrez Gene ID: (Human) 5547, (Mouse) 72461
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support