Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IITTDENECN WMMFVRKARN REEQNLVAYP HDGKIFFCTS QDIPPENELL FYYSRDYAQQ IGVPEHPDVH LCNCGKECNS YTEFKAHLTS HIHNHLPTQG
The protein encoded by this gene is a transcription factor of the PR-domain protein family. It contains a PR-domain and multiple zinc finger motifs. Transcription factors of the PR-domain family are known to be involved in cell differentiation and tumorigenesis. An elevated expression level of this gene has been observed in PC12 cells treated with nerve growth factor, beta polypeptide (NGF). This gene is located in a chromosomal region that is thought to contain tumor suppressor genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC45046; PR domain containing 4; PR domain zinc finger protein 4; PR domain-containing protein 4; PR-domain zinc-finger protein PFM1
Gene Aliases: PFM1; PRDM4
UniProt ID: (Human) Q9UKN5
Entrez Gene ID: (Human) 11108
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support