Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: AKGDSESEED EDLEVPVPSR FNRRVSVCAE TYNPDEEEED TDPRVIHPKT DEQRCRLQEA CKDILLFKNL DQEQLSQVLD AMFERIVKAD EHVIDQGDDG DNFYVIERGT YDILVTKDNQ TRSVGQYDNR GSFGELALMY NTPRAATIVA TSEGSLWGLD RVTFRRIIVK NNAKKRKMFE SFIESVPLLK SLEVSERMKI VDVIGEKIYK DGERIITQTK SNKDGGNQEV EIARCHKGQY FGELALVTNK PRAASAYAVG DVKCLVMDVQ AFERLLGPCM DIMKRNISHY EEQLVKMFGS SVDLGNLGQ
cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP-dependent protein kinase (AMPK), which transduces the signal through phosphorylation of different target proteins. The inactive holoenzyme of AMPK is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits of AMPK have been identified in humans. The protein encoded by this gene is one of the regulatory subunits. This subunit can be phosphorylated by the activated catalytic subunit. It may interact with various A-kinase anchoring proteins and determine the subcellular localization of AMPK.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cAMP-dependent protein kinase regulatory subunit RII alpha; cAMP-dependent protein kinase type II-alpha regulatory subunit; MGC3606; protein kinase A, RII-alpha subunit; protein kinase cAMP-dependent regulatory type II alpha; protein kinase, cAMP-dependent, regulatory subunit type II alpha; protein kinase, cAMP-dependent, regulatory, type 2, alpha
Gene Aliases: 1110061A24Rik; AI317181; AI836829; PKR2; PRKAR2; PRKAR2A; RII(alpha)
UniProt ID: (Human) P13861, (Rat) P12368
Entrez Gene ID: (Human) 5576, (Mouse) 19087, (Rat) 29699
Molecular Function: protein-binding activity modulator kinase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support