Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive Samples: Mouse brian, Rat brain
Immunogen sequence: GEVKITMKRK GKSEVEENGA VEIARCSRGQ YFGELALVTN KPRAASAHAI GTVKCLAMDV QAFERLLGPC MEIMKRNIAT YEEQLVALFG TNMDIVEPTA
cAMP is a signaling molecule important for a variety of cellular functions. cAMP exerts its effects by activating the cAMP-dependent protein kinase, which transduces the signal through phosphorylation of different target proteins. The inactive kinase holoenzyme is a tetramer composed of two regulatory and two catalytic subunits. cAMP causes the dissociation of the inactive holoenzyme into a dimer of regulatory subunits bound to four cAMP and two free monomeric catalytic subunits. Four different regulatory subunits and three catalytic subunits have been identified in humans. The protein encoded by this gene is one of the regulatory subunits. This subunit can be phosphorylated by the activated catalytic subunit. This subunit has been shown to interact with and suppress the transcriptional activity of the cAMP responsive element binding protein 1 (CREB1) in activated T cells. Knockout studies in mice suggest that this subunit may play an important role in regulating energy balance and adiposity. The studies also suggest that this subunit may mediate the gene induction and cataleptic behavior induced by haloperidol.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cAMP-dependent protein kinase type II-beta regulatory subunit; Type II beta regulatory subunit of cAMP-dependent protein kinase
Gene Aliases: AI451071; AW061005; Pkarb2; PKARIIbeta; Prkar2b; RATDNA; RII(beta)
UniProt ID: (Mouse) P31324, (Rat) P12369
Entrez Gene ID: (Mouse) 19088, (Rat) 24679
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support