Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LPICPGGAAR CQVTLRDLFD RAVVLSHYIH NLSSEMFSEF DKRYTHGRGF ITKAINSCHT SSLATPEDKE QAQQMNQKDF LSLIVSILRS WNEPLYHLVT EVRGMQEAPE AILSKAVEIE EQTKRLLEGM ELIVSQVHPE TKENEIYPVW SGLPSLQMAD EESRLSAYYN LLHCLRRDSH KIDNYLKLLK CRIIHNNNC
Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: decidual prolactin; growth hormone A1; Human prolactin; PRL; Prolactin
Gene Aliases: GHA1; PRL
UniProt ID: (Human) P01236
Entrez Gene ID: (Human) 5617
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support