Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MARYRCCRSQ SRSRYYRQRQ RSRRRRRRSC QTRRRAMRCC RPRYRPRCRR H
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen family 94, member 1; Cysteine-rich protamine; Sperm protamine P1; testicular tissue protein Li 91; testis specific protamine 1
Gene Aliases: CT94.1; P1; PRM1
UniProt ID: (Human) P04553
Entrez Gene ID: (Human) 5619
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support