Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RAREDLQKM KQFAQDFVMH TDVRTCSSST SVIADLQEDE DGVYFSSYGH YGIHEEMLKD KIRTESYRDF IYQNPHIFKD KVVLDVGCGT GILSMFAAKA GAKKVLGVDQ SEILYQAMDI IRLNKLEDTI TLIKGKIEEV HLPVEKVDVI ISEWMGYFLL FESMLDSVLY AKNKYLAKGG SVYPDICTIS LVAVSDVNKH ADRIAFWDDV YGFKMSCMKK AVIPEAVVEV LDPKTLISEP CGIKHIDCHT TSISDLEFSS DFTLKITRTS MCTAIAGYFD IYFEKNCHNR VVFSTGPQST KTHWKQTVFL LEKPFSVKAG EALKGKVTVH KNKKDPRSLT VTLTLNNSTQ TYGLQ (178-531 aa encoded by BC037544)
Type I protein arginine N-methyltransferases (PRMTs), such as PRMT3, catalyze the formation of asymmetric N(G),N(G)-dimethylarginine (ADMA) residues in proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: heterogeneous nuclear ribonucleoprotein methyltransferase-like 3; Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3; HMT1 hnRNP methyltransferase-like 3; protein arginine methyltransferase 3; Protein arginine N-methyltransferase 3; protein arginine N-methyltransferase 3(hnRNP methyltransferase S. cerevisiae)-like 3
Gene Aliases: 2010005E20Rik; 2410018A17Rik; AL033309; HRMT1L3; PRMT3
UniProt ID: (Human) O60678, (Rat) O70467, (Mouse) Q922H1
Entrez Gene ID: (Human) 10196, (Rat) 89820, (Mouse) 71974
Molecular Function:
protein modifying enzyme
zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support