Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Specificity: This antibody recognizes PRMT3 (Protein arginine N-methyltransferase 3). It binds to a folded domain, amino acids 211-531. PRMT3 methylates (mono and asymmetric dimethylation) the guanidino nitrogens of arginyl residues in some proteins.
Type I protein arginine N-methyltransferases (PRMTs), such as PRMT3, catalyze the formation of asymmetric N(G),N(G)-dimethylarginine (ADMA) residues in proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 3; HMT1 hnRNP methyltransferase-like 3; Protein arginine N-methyltransferase 3
Gene Aliases: HRMT1L3; PRMT3
UniProt ID: (Human) O60678
Entrez Gene ID: (Human) 10196
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support