Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LVRGAYLAQE RARAAEIGYE DPINPTYEAT NAMYHRCLDY VLEELKHNAK AKVMVASHNE DTVRFALRRM EELGLHPADH RVYFGQLLGM CDQISFPLGQ
The protein encoded by this gene is a mitochondrial proline dehydrogenase that catalyzes the first step in proline degradation. Defects in this gene are a cause of hyperprolinemia type 1 and possibly susceptibility to schizophrenia 4 (SCZD4). The gene is located on chromosome 22q11.21, a region which has also been associated with the contiguous gene deletion syndromes DiGeorge syndrome and CATCH22 syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: p53-induced gene 6 protein; PRODH 1; PRODH 2; proline dehydrogenase (oxidase) 1; Proline dehydrogenase 1, mitochondrial; Proline oxidase; Proline oxidase 2; proline oxidase, mitochondrial; SCZD4; tumor protein p53 inducible protein 6
Gene Aliases: HSPOX2; PIG6; POX; POX2; PRODH; PRODH1; PRODH2; TP53I6
UniProt ID: (Human) O43272
Entrez Gene ID: (Human) 5625
Molecular Function:
oxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support