Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive test controls include: 293T, HepG2, Mouse liver, Mouse kidney, Rat ovary, Rat kidney.
Immunogen sequence: MRGATRVSIM LLLVTVSDCA VITGACERDV QCGAGTCCAI SLWLRGLRMC TPLGREGEEC HPGSHKVPFF RKRKHHTCPC LPNLLCSRFP DGRYRCSMDL KNINF
Endocrine gland-derived vascular endothelial growth factor induces proliferation, migration, and fenestration in capillary endothelial cells derived from endocrine glands. Its expression is induced by hypoxia and is restricted to the steroidogenic glands. Its expression is often complementary to the expression of VEGF, suggesting that these molecules function in a coordinated manner.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: black mamba toxin-related protein; EG-VEGF; Endocrine-gland-derived vascular endothelial growth factor; Mambakine; prok1; Prokineticin-1
Gene Aliases: EG-VEGF; EGVEGF; PK1; PRK1; PROK1; UNQ600/PRO1186
UniProt ID: (Human) P58294, (Mouse) Q14A28, (Rat) Q8R414
Entrez Gene ID: (Human) 84432, (Mouse) 246691, (Rat) 192205
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support