Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Catalyzes the synthesis of phosphoribosylpyrophosphate (PRPP) that is essential for nucleotide synthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hCG_1643027; Phosphoribosyl pyrophosphate synthase 1-like 1; Phosphoribosyl pyrophosphate synthase III; phosphoribosylpyrophosphate synthetase subunit III; PRPS1-like 1; PRS-III; ribose-phosphate diphosphokinase catalytic chain III; Ribose-phosphate pyrophosphokinase 3; ribose-phosphate pyrophosphokinase III
Gene Aliases: 1700011K15Rik; PRPS1; PRPS1L1; PRPS3; PRPSL; PRS-III
UniProt ID: (Human) P21108
Entrez Gene ID: (Human) 221823, (Rat) 298954, (Mouse) 75456
Molecular Function:
nucleotide kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support