Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: AEWKNCIIVS PDAGGAKRVT SIADRLNVEF ALIHKERKKA NEVDRMVLVG DVKDRVAILV DDMADTCGTI CHAADKLLSA GATKVYAILT HGIFSGPAIS RINNAAFEAV
The function remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Phosphoribosyl pyrophosphate synthase II; PPRibP; PPRibP synthetase; PRS-II; ribose-phosphate diphosphokinase 2; Ribose-phosphate pyrophosphokinase 2; testicular secretory protein Li 41
Gene Aliases: PRPS2; PRSII
UniProt ID: (Human) P11908
Entrez Gene ID: (Human) 5634
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support