Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: WRCFLRNKT RRQTVTEGHI PFPQHLNIIT PPPPPDEVFD SSGLSPGFLG YVVGRSDSVS TRLSNCDPPP TYEEATGQVN LQRSETEPHL DPPPEYEDIV NSNSASAIPM VPVVTTIK (102-218 aa encoded by BC060833)
This gene encodes a vitamin K-dependent, gamma-carboxyglutamic acid (Gla)-containing, single-pass transmembrane protein. This protein contains a Gla domain at the N-terminus, preceded by a propeptide sequence required for post-translational gamma-carboxylation of specific glutamic acid residues by a vitamin K-dependent gamma-carboxylase. The C-terminus is proline-rich containing PPXY and PXXP motifs found in a variety of signaling and cytoskeletal proteins. This gene is highly expressed in the spinal cord. Several alternatively spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ortholog of proline rich Gla (G-carboxyglutamic acid) 1 PRRG1; proline rich Gla (G-carboxyglutamic acid) 1; Proline-rich gamma-carboxyglutamic acid protein 1; proline-rich Gla (G-carboxyglutamic acid) polypeptide 1; Proline-rich Gla protein 1; Transmembrane gamma-carboxyglutamic acid protein 1
Gene Aliases: 2010007L08Rik; PRGP1; PRRG1; RGD1561320; TMG1
UniProt ID: (Human) O14668
Entrez Gene ID: (Human) 5638, (Mouse) 546336, (Rat) 363472
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support