Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: VSNIYLSPRY LGNSPYDIAL VKLSAPVTYT KHIQPICLQA STFEFENRTD CWVTGWGYIK EDEALPSPHT LQEVQVAIIN NSMCNHLFLK YSFRKDIFGD
Could regulate proteolytic events associated with testicular germ cell maturation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 3.4.21; Eosinophil serine protease 1; ESP-1; protease, serine, 21 (testisin); Serine protease 21; serine protease from eosinophils; TEST1testisin; Testisin
Gene Aliases: ESP-1; ESP1; PRSS21; TEST1; TESTISIN; UNQ266/PRO303
UniProt ID: (Human) Q9Y6M0
Entrez Gene ID: (Human) 10942
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support