Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RALPAEYRVR LGALRLGSTS PRTLSVPVRR VLLPPDYSED GARGDLALLQ LR
Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 73%.
Serine protease that has amidolytic activity, cleaving its substrates before Arg residues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: protease, serine, 33; Serine protease 33; Serine protease EOS
Gene Aliases: EOS; PRSS33
UniProt ID: (Human) Q8NF86
Entrez Gene ID: (Human) 260429
Molecular Function:
serine protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support