Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total PSAP.
PSAP is a highly conserved glycoprotein which is a precursor for 4 cleavage products: saposins A, B, C, and D. Each domain of the precursor protein is approximately 80 amino acid residues long with nearly identical placement of cysteine residues and glycosylation sites. Saposins A-D localize primarily to the lysosomal compartment where they facilitate the catabolism of glycosphingolipids with short oligosaccharide groups. The precursor protein exists both as a secretory protein and as an integral membrane protein and has neurotrophic activities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Proactivator polypeptide; Prosaposin; prosaposin (sulfated glycoprotein, sphingolipid hydrolase activator); SGP-1; snoRNA MBII-198; sphingolipid activator protein-1; Sulfated glycoprotein 1
Gene Aliases: AI037048; GLBA; PSAP; SAP1; SGP-1; Sgp1; SGP1A
UniProt ID: (Human) P07602, (Rat) P10960, (Mouse) Q61207
Entrez Gene ID: (Human) 5660, (Rat) 25524, (Mouse) 19156
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support