Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody is specific to PSAPL1.
May activate the lysosomal degradation of sphingolipids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Proactivator polypeptide-like 1; prosaposin-like protein 1
Gene Aliases: PSAPL1
UniProt ID: (Human) Q6NUJ1
Entrez Gene ID: (Human) 768239
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support