Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDAPRQVVNF GPGPAKLPHS VLLEIQKELL DYKGVGISVL EMSHRSSDFA KIINNTENLV RELLAVPDNY KVIFLQGGGC GQFSAVPLNL IGLKAGRCAD YVVTGAWSAK AAEEAKKFGT INIVHPKLGS YTKIPDPSTW NLNPDASYVY YCANETVHGV EFDFIPDVKG AVLVCDMSSN FLSKPVDVSK FGVIFAGAQK NVGSAGVTVV IVRDDLLGFA LRECPSVLEY KVQAGNSSLY NTPPCFSIYV MGLVLEWIKN NGGAAAMEKL SSIKSQTIYE IIDNSQGFYV CPVEPQNRSK MNIPFRIGNA KGDDALEKRF LDKALELNML SLKGHRSVGG IRASLYNAVT IEDVQKLAAF MKKFLEMHQL
The protein encoded by this gene is likely a phosphoserine aminotransferase, based on similarity to proteins in mouse, rabbit, and Drosophila. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Endometrial progesterone-induced protein; EPIP; Phosphohydroxythreonine aminotransferase; Phosphoserine aminotransferase; PSAT
Gene Aliases: D8Ertd814e; EPIP; NLS2; PSA; PSAT; PSAT1; PSATD
UniProt ID: (Human) Q9Y617, (Mouse) Q99K85
Entrez Gene ID: (Human) 29968, (Pig) 100154160, (Mouse) 107272
Molecular Function:
transaminase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support