Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: CYSCKAQVSN EDCLQVENCT QLGEQCWTAR IRAVGLLTVI SKGCSLNCVD DSQDYYVGKK NITCCDTDLC NAS
PSCA (Prostate Stem Cell antigen) is a cell surface antigen, which is overexpressed in approximately40% of primary prostate cancers and in as many as 100% of metastatic ones. PSCA is also overexpressed in a majority of transitional cell and pancreatic carcinomas. Antibody directed against PSCA inhibits tumorigenesis, slows tumor growth, prolongs survival and prevents metastasis in a preclinical zenograft model indicating that PSCA may have utility as a prognostic marker and/or therapeutic target in prostate cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: caldesmon; H-CAD; HCAD; L-CAD; NAG22; Prostate stem cell antigen; UNQ206
Gene Aliases: PRO232; PSCA; UNQ206/PRO232
UniProt ID: (Human) O43653
Entrez Gene ID: (Human) 8000
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support