Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MHAAEIMGPL SAPPCTEHIK WKGLLLTVET PKPSISSSNL NPREAMETVI LTCNPETPDA SYLWWMNGQS LPMTHRMQLS ETNRTLFLFG VTKYTAGPYE CEIWNSGSAS RSDPVTLNLL HGPDLPRIFP SVTSYYSGEN LDLSCFADSN PPAQYSWTIN GKFQLSGQKL FIPQITPKHN GLYACSARNS ATGEESSTSL TIRVIAPPGL GTFAFNNPT
PSG11, also named as PSG13, PSG14, PSBG-11 and PSBG-13, belongs to the immunoglobulin superfamily and CEA family. PSBG are produced in high quantity during pregnancy. PSG11 and other PSGs induce dose-dependent secretion of anti-inflammatory cytokines by human monocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Pregnancy-specific beta-1-glycoprotein 11; Pregnancy-specific beta-1-glycoprotein 13; PS-beta-G-11; PS-beta-G-13
Gene Aliases: PSBG-11; PSBG-13; PSG11; PSG13; PSG14
UniProt ID: (Human) Q9UQ72
Entrez Gene ID: (Human) 5680
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support