Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: YTLHIIKGD DGTRGVTGRF TFTLHLETPK PSISSSNLNP RETMEAVSLT CDPETPDASY LWWMNGQSLP MTHSLKLSET NRTLFLLGVT KYTAGPYECE IRNPVSASRS DPVTLNLLPK LSKPYITINN LNPRENKDVL TFTCEPKSEN YTYIWWLNGQ SLPVSPRVKR PIENRILILP NVTRNETGPY QCEIRDRYGG IRSDPVTLNV LYGPDLPSIY PSFTYYRSGE NLYLSCFAES NPRAQYSWTI NGKFQLSGQK LSIPQITTKH SGLYACSVRN SATGKESSKS ITVKVSDWIL P (120-419 aa encoded by BC008405)
The protein encoded by this gene is a pregnancy-specific glycoprotein (PSG), one of several encoded by a cluster of similar genes on chromosome 19. This gene is a member of the carcinoembryonic antigen (CEA) gene family and may play a role in regulation of the innate immune system. Several transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Pregnancy-specific beta-1-glycoprotein 4; Pregnancy-specific beta-1-glycoprotein 9; Pregnancy-specific beta-1-glycoprotein-4; pregnancy-specific glycoprotein 4; pregnancy-specific glycoprotein 9; PS-beta-G-4; PS-beta-G-9
Gene Aliases: CGM4; PSBG-4; PSBG-9; PSG4; PSG9
UniProt ID: (Human) Q00888
Entrez Gene ID: (Human) 5672
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support