Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAERGYSFS LTTFSPSGKL VQIEYALAAV AGGAPSVGIK AANGVVLATE KKQKSILYDE RSVHKVEPIT KHIGLVYSGM GPDYRVLVHR ARKLAQQYYL VYQEPIPTAQ LVQRVASVMQ EYTQSGGVRP FGVSLLICGW NEGRPYLFQS DPSGAYFAWK ATAMGKNYVN GKTFLEKRYN EDLELEDAIH TAILTLKESF EGQMTEDNIE VGICNEAGFR RLTPTEVKDY LAAIA (1-234 aa encoded by BC047697)
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the peptidase T1A family, that is a 20S core alpha subunit.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Macropain subunit C3; Multicatalytic endopeptidase complex subunit C3; OTTHUMP00000207364; OTTHUMP00000207366; proteasome (prosome, macropain) subunit, alpha type, 2; proteasome alpha 2 subunit; Proteasome component C3; Proteasome subunit alpha type-2; proteasome subunit HC3; PSMA 2
Gene Aliases: HC3; Lmpc3; MU; PMSA2; PSC2; PSC3; PSMA2
UniProt ID: (Human) P25787, (Rat) P17220, (Mouse) P49722
Entrez Gene ID: (Human) 5683, (Rat) 29669, (Mouse) 19166
Molecular Function:
protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support