Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MALASVLERP LPVNQRGFFG LGGRADLLDL GPGSLSDGLS LAAPGWGVPE EPGIEMLHGT TTLAFKFRHG VIVAADSRAT AGAYIASQTV KKVIEINPYL LGTMAGGAAD CSFWERLLAR QCRIYELRNK ERISVAAASK LLANMVYQYK GMGLSMGTMI CGWDKRGPGL YYVDSEGNRI SGATFSVGSG SVYAYGVMDR GYSYDLEVEQ AYDLARRAIY QATYRDAYSG GAVNLYHVRE DGWIRVSSDN VADLHEKYSG STP
Cyclic nucleotide phosphodiesterases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Macropain epsilon chain; MGC118075; Multicatalytic endopeptidase complex epsilon chain; proteasome (prosome, macropain) subunit, beta type, 5; proteasome beta 5 subunit; proteasome catalytic subunit 3; Proteasome chain 6; Proteasome epsilon chain; Proteasome subunit beta type-5; Proteasome subunit MB1; Proteasome subunit X; proteasome subunit, beta type, 5; PSX large multifunctional protease X; testicular tissue protein Li 153
Gene Aliases: LMPX; MB1; PSMB5; X
UniProt ID: (Human) P28074
Entrez Gene ID: (Human) 5693
Molecular Function: protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support