Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MPDYLGADQR KTKEDEKDDK PIRALDEGDI ALLKTYGQST YSRQIKQVED DIQQLLKKIN ELTGIKESDT GLAPPALWDL AADKQTLQSE QPLQVARCTK IINADSEDPK YIINVKQFAK FVVDLSDQVA PTDIEEGMRV GVDRNKYQIH IPLPPKIDPT VTMMQVEEKP DVTYSDVGGC KEQIEKLREV VETPLLHPER FVNLGIEPPK GVLLFGPPGT GKTLCARAVA NRTDACFIRV IGSELVQKYV GEGARMVREL FEMARTKKAC LIFFDEIDAI GGARFDDGAG GDNEVQRTML ELINQLDGFD PRGNIKVLMA TNRPDTLDPA LMRPGRLDRK IEFSLPDLEG RTHIFKIHAR SMSVERDIRF ELLARLCPNS TGAEIRSVCT EAGMFAIRAR RKIATEKDFL EAVNKVIKSY AKFSATPRYM TYN
Proteolytic degradation is critical to the maintenance of appropriate levels of short-lived and regulatory proteins as important and diverse as those involved in cellular metabolism, heat shock and stress response, antigen presentation, modulation of cell surface receptors and ion channels, cell cycle regulation, transcription, and signalling factors. The ubiquitin-proteasome pathway deconstructs most proteins in the eukaryotic cell cytosol and nucleus. Others are degraded via the vacuolar pathway which includes endosomes, lysosomes, and the endoplasmic reticulum. The 26S proteasome is an ATP-dependent, multisubunit (~31), barrel-shaped molecular machine with an apparent molecular weight of ~2.5 MDa. It consists of a 20S proteolytic core complex which is crowned at one or both ends by 19S regulatory subunit complexes. The 19S regulatory subunits recognize ubiquitinated proteins and play an essential role in unfolding and translocating targets into the lumen of the 20S subunit. An enzymatic cascade is responsible for the attachment of multiple ubiquitin molecules to lysine residues of proteins targeted for degradation. Several genetic diseases are associated with defects in the ubiquitin-proteasome pathway. Some examples of affected proteins include those linked to cystic fibrosis, Angelman’s syndrome, and Liddle syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 26S proteasome AAA-ATPase subunit RPT1; 26S proteasome regulatory subunit 7; mammalian suppressor of sgv-1 of yeast; MGC3004; protease 26S subunit 7; proteasome (prosome, macropain) 26S subunit, ATPase, 2; Proteasome 26S subunit ATPase 2; proteasome 26S subunit, ATPase, 2; putative protein product of Nbla10058; testis secretory sperm-binding protein Li 197a
Gene Aliases: MSS1; Nbla10058; PSMC2; S7
UniProt ID: (Human) P35998
Entrez Gene ID: (Human) 5701
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support