Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Positive control: Hela cell lysate, 293T cell lysate, SH-SY5Y cell lysate, HepG2 cell lysate, PC-12 cell lysate, NIH/3T3 cell lysate, mouse kidney tissue lysate, rat kidney tissue lysate, rat brain tissue lysate, human testis tissue, SH-SY5Y, NIH/3T3, PC-12.
Predicted band size: 44 kDa
Subcellular Location: Cytoplasm, Nucleus.
PSMC6 (proteasome (prosome, macropain) 26S subunit, ATPase 6), also known as P44, p42, SUG2, S10B or CADP44, is a regulatory component of the 26S proteasome. More specifically, PSMC6 is one of the six ATPase subunits of the 19S regulator base. It contains a leucine zipper motif and an AAA (ATPase associated with diverse cellular activities) domain and belongs to the AAA ATPase family of chaperone-like ATPases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 26S protease regulatory subunit 10B; 26S protease regulatory subunit S10B; 26S proteasome AAA-ATPase subunit RPT4; 26S proteasome regulatory subunit 10B; conserved ATPase domain protein 44; epididymis secretory protein Li 73; proteasome (prosome, macropain) 26S subunit, ATPase, 6; proteasome (prosome, macropain) 26S subunit, ATPase, 6-like; Proteasome 26S subunit ATPase 6; Proteasome subunit p42
Gene Aliases: 2300001E01Rik; AI451058; CADP44; HEL-S-73; p42; P44; PSMC6; SUG2
UniProt ID: (Human) P62333, (Mouse) P62334
Entrez Gene ID: (Human) 5706, (Mouse) 67089, (Rat) 289990
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support