Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Aliquoting is unnecessary for -20°C storage.
Acts as a regulatory subunit of the 26S proteasome which is involved in the ATP-dependent degradation of ubiquitinated proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 26S proteasome non-ATPase regulatory subunit 13; 26S proteasome non-ATPase regulatory subunit 13 {ECO:0000250|UniProtKB:Q9UNM6, ECO:0000312|EMBL:AAI58733.1}; 26S proteasome regulatory subunit p40.5; 26S proteasome regulatory subunit p40.5 {ECO:0000250|UniProtKB:Q9UNM6}; 26S proteasome regulatory subunit RPN9; 26S proteasome regulatory subunit S11; 26S proteasome regulatory subunit S11 {ECO:0000250|UniProtKB:Q9UNM6}; 26S proteasome subunit p40.5; proteasome (prosome, macropain) 26S subunit, non-ATPase, 13; psmd13 {ECO:0000312|EMBL:AAI58733.1, ECO:0000312|RGD:1305236}
Gene Aliases: HSPC027; p40.5; PSMD13; Rpn9; S11
UniProt ID: (Human) Q9UNM6, (Mouse) Q9WVJ2, (Rat) B0BN93
Entrez Gene ID: (Human) 5719, (Mouse) 23997, (Rat) 365388
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support