Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
The 26S proteasome is responsible for most intracellular proteolysis in eukaryotes. POH1 (also known as Rpn11 in yeast) subunit of the 19S complex is responsible for substrate deubiquitination during proteasomal degradation. POH1 appears to be a Zn21-dependent protease, unlike all other known deubiquitinating enzymes (DUBs) that are cysteine proteases. POH1 was first identified as the human functional homolog of the S. pombe pad1 gene, which confers moderate resistance to chemotherapeutic drugs, and also to ultraviolet light when transiently overproduced in mammalian cells. Poh1-induced drug resistance is mediated through AP-1 transcription factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 26S proteasome non-ATPase regulatory subunit 14; 26S proteasome non-ATPase subunit; 26S proteasome regulatory subunit RPN11; 26S proteasome-associated pad1 homolog; 26S proteasome-associated PAD1 homolog 1; MAD1; proteasome (prosome, macropain) 26S subunit, non-ATPase, 14; proteasome 26S subunit, non-ATPase 14; testis tissue sperm-binding protein Li 69n
Gene Aliases: 2610312C03Rik; 3200001M20Rik; AA986732; PAD1; POH1; PSMD14; rpm11; RPN11
UniProt ID: (Human) O00487, (Mouse) O35593
Entrez Gene ID: (Human) 10213, (Rat) 311078, (Mouse) 59029
Molecular Function:
translation initiation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support