Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to PSMD5.
Immunogen sequence: LTQAGLEAL FESNLLDDLK SVMKTNDIVR YRVYELIIEI SSVSPESLNY CTTSGLVTQL LRELTGEDVL VRATCIEMVT SLAYTHHGRQ YLAQEGVIDQ ISNIIVGADS DPFSSFYLPG FVKFFGNLAV MDSPQQICER YPIFVEKVFE MIESQDPTMI GVAVDTVGIL GSNVEGKQVL QKTGTRFERL LMRIGHQSKN APVELKIRCL DAISSLLYLP PEQQTDDLLR MTESWFSSLS RDPLELFRGI SSQPFPELHC AALKVFTAIA NQPWAQKLMF NSPGFVEYVV DRSVEHDKAS KDAKYELVKA LANSKTIAEI FGNPNYLRLR TYLSEGPYYV KPVSTTAVEG AE (154-504 aa encoded by BC014478)
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a non-ATPase subunit of the 19S regulator base.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 26S protease subunit S5 basic; 26S proteasome non-ATPase regulatory subunit 5; 26S proteasome subunit S5B; MGC23145; proteasome (prosome, macropain) 26S subunit, non-ATPase, 5; RP11-27I1.5
Gene Aliases: 1500032A03Rik; AW475925; KIAA0072; mKIAA0072; PSMD5; S5B; W91691
UniProt ID: (Human) Q16401, (Mouse) Q8BJY1
Entrez Gene ID: (Human) 5711, (Mouse) 66998, (Rat) 296651
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support