Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MAAQALALLR EVARLEAPLE ELRALHSVLQ AVPLNELRQQ AAELRLGPLF SLLNENHREK TTLCVSILER LLQAMEPVHV ARNLRVDLQR GLIHPDDSVK ILTLSQIGRI VENSDAVTEI LNNAELLKQI VYCIGGENLS VAKAAIKSLS RISLTQAGLE ALFESNLLDD LKSVMKTNDI VRYRVYELII EISSVSPESL NYCTTSGLVT QLLRELTGED VLVRATCIEM VTSLAYTHHG RQYLAQEGVI DQISNIIVGA DSDPFSSFYL PGFVKFFGNL AVMDSPQQIC ERYPIFVEKV FEMIESQDPT MIGVAVDTVG ILGSNVEGKQ VLQKTGTRFE RLLMRIGHQS KNAPVELKIR CLDAISSLLY LPPEQQTDDL LRMTESWFSS LSRDPLELFR GISSQPFPEL HCAALKVFTA IANQPWAQKL MFNSPGFVEY VVDRSVEHDK ASKDAKYELV KALANSKTIA EIFGNPNYLR LRTYLSEGPY YVKPVSTTAV EGAE
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a non-ATPase subunit of the 19S regulator base.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 26S protease subunit S5 basic; 26S proteasome non-ATPase regulatory subunit 5; 26S proteasome subunit S5B; MGC23145; proteasome (prosome, macropain) 26S subunit, non-ATPase, 5; RP11-27I1.5
Gene Aliases: KIAA0072; PSMD5; S5B
UniProt ID: (Human) Q16401
Entrez Gene ID: (Human) 5711
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support