Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the protease subunit S10 family. The encoded protein is a subunit of the 26S proteasome which colocalizes with DNA damage foci and is involved in the ATP-dependent degradation of ubiquinated proteins. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 26S proteasome non-ATPase regulatory subunit 6; 26S proteasome regulatory subunit RPN7; 26S proteasome regulatory subunit S10; Breast cancer-associated protein SGA-113M; p42A; Phosphonoformate immuno-associated protein 4; proteasome (prosome, macropain) 26S subunit, non-ATPase, 6; Proteasome regulatory particle subunit p44S10; proteasome, 26S, non-ATPase regulatory subunit 6
Gene Aliases: 2400006A19Rik; KIAA0107; p42A; P44s10; PFAAP4; PSMD6; Rpn7; S10; SGA-113M
UniProt ID: (Human) Q15008, (Mouse) Q99JI4
Entrez Gene ID: (Human) 9861, (Rat) 289924, (Mouse) 66413
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support