Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSDEEARQSG GSSQAGVVTV SDVQELMRRK EEIEAQIKAN YDVLESQKGI GMNEPLVDCE GYPRSDVDLY QVRTARHNII CLQNDHKAVM KQVEEALHQL HA
Acts as a chaperone during the assembly of the 26S proteasome, specifically of the base subcomplex of the PA700/19S regulatory complex (RC). During the base subcomplex assembly is part of an intermediate PSMD9:PSMC6:PSMC3 module, also known as modulator trimer complex; PSMD9 is released during the further base assembly process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 26S proteasome non-ATPase regulatory subunit 9; 26S proteasome regulatory subunit p27; homolog of rat Bridge 1; MGC8644; proteasome (prosome, macropain) 26S subunit, non-ATPase, 9; proteasome 26S non-ATPase regulatory subunit 9; Transactivating protein Bridge-1
Gene Aliases: 1500011J20Rik; 2610202L11Rik; Bridge; Bridge-1; P27; PSMD9; Rpn4
UniProt ID: (Human) O00233, (Mouse) Q9CR00, (Rat) Q9WTV5
Entrez Gene ID: (Human) 5715, (Mouse) 67151, (Rat) 161475
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support