Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CCPLLEQPNI VHDLPAAVLS YCQVWKIPAI LYLCYTDVMK LDLITVEAFK PILSTRSLKG LVKNIPQSTE ILKKLMTTNE IQSNIYT
Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 87%.
Chaperone protein which promotes assembly of the 20S proteasome as part of a heterodimer with PSMG2. The PSMG1-PSMG2 heterodimer binds to the PSMA5 and PSMA7 proteasome subunits, promotes assembly of the proteasome alpha subunits into the heteroheptameric alpha ring and prevents alpha ring dimerization.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C21-LRP; Chromosome 21 leucine-rich protein; Down syndrome critical region gene 2; Down syndrome critical region protein 2; leucine rich protein C21-LRP; PAC-1; proteasome (prosome, macropain) assembly chaperone 1; proteasome assembling chaperone 1; Proteasome assembly chaperone 1
Gene Aliases: C21LRP; DSCR2; LRPC21; PAC-1; PAC1; PSMG1
UniProt ID: (Human) O95456
Entrez Gene ID: (Human) 8624
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support