Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Immunogen sequence: MDGIVTEVAV GVKRGSDELL SGSVLSSPNS NMSSMVVTAN GNDSKKFKGE DKMDGAPSRV LHIRKLPGEV TETEVIALGL PFGKVTNILM LKGKNQAFLE LATEEAAITM VNYYSAVTPH LRNQPIYIQY SNHKELKTDN TLNQRAQAVL QAVTAVQTAN TPLSGTTVSE SAVTPAQSPV LRIIIDNMYY PVTLDVLHQI FSKFGAVLKI ITFTKNNQFQ ALLQYGDPVN AQQAKLALDG QNIYNACCTL RIDFSKLVNL NVKYNNDKSR DYTRPDLPSG; Positive Samples: SH-SY5Y, HeLa, Mouse brain, Mouse testis, Rat testis; Cellular Location: Nucleus
PTBP2 encodes a protein that binds to the intronic cluster of RNA regulatory elements, downstream control sequence (DCS). It is implicated in controlling the assembly of other splicing-regulatory proteins. This protein is very similar to the polypyrimidine tract binding protein but it is expressed primarily in the brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: br n nPTB5; Brain-enriched polypyrimidine tract-binding protein; Brain-enriched PTB; neural polypyrimidine tract binding protein; Neural polypyrimidine tract-binding protein; Neurally-enriched homolog of PTB; nPTB5; nPTB6; nPTB7; nPTB8; Polypyrimidine tract-binding protein 2; PTB-like protein; RRM-type RNA-binding protein brPTB
Gene Aliases: Brptb; NPTB; PTB; Ptb2; PTBLP; PTBP2
UniProt ID: (Human) Q9UKA9, (Mouse) Q91Z31, (Rat) Q66H20
Entrez Gene ID: (Human) 58155, (Mouse) 56195, (Rat) 310820
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support