Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Antibody detects endogenous levels of total PTCH2.
Mutations of the human Patched tumor suppressor gene (PTCH) have been identified in individuals with the nevoid basal cell carcinoma syndrome (NBCCS) as well as in sporadic basal cell carcinomas and medulloblastomas. PTCH2 has a frameshift mutation resulting in a truncated PTCH2 protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: patched homolog 1; patched homolog 2; Protein patched homolog 2; PTC2; RP11-269F19.8
Gene Aliases: PTC2; Ptch1; PTCH2; UNQ560/PRO1121/PRO57079
UniProt ID: (Human) Q9Y6C5, (Mouse) O35595
Entrez Gene ID: (Human) 8643, (Mouse) 19207, (Rat) 366452
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support